Glutaraldehyde 25% in aqueous solution for electron microscopy
250 mL
Supplier: VWR Chemicals
Glutaraldehyde 25% in aqueous solution for electron microscopy
Catalog # 87043.230
Supplier: VWR Chemicals
CAS Number: 111-30-8
Jump to:
Product Details & Documents
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
- CAS Number:111-30-8
- Formula:C₅H₈O₂
- MDL Number:MFCD00007025
- Molecular Weight:100.12 g/mol
- Melting Pt:–6 °C
- Storage Temperature:Ambient
- UN:2922
- ADR:8 (6.1),II
- :Density: 1.014 g/cm³ (20 °C)
Documents
Certificates
Search by Lot Numberspec_87043.230.pdf spec_87043.290 spec_87043.320
Product specification
Specifications
Specification Test Results
- Appearance:clear colorless or yellow liquid
- Assay :23,5 – 26,5%
- Color scale:max. 50 APHA
- pH:min. 3,0
- Density (d20°C/4°C):1,060 - 1,065
- Absorbance A (235 nm; 1:200 diluted with water; 1 cm):max. 0,6
Specifications
- Pk:250 mL



