Orders LinkContactUsLinkComponent
Clips for conical ground joints
Clips for conical ground joints
Green, 24/29
Catalog # HECH42386024
Clips for conical ground joints
Catalog # HECH42386024
Clips for conical ground joints
Clips for conical ground joints
Green, 24/29
Price
31,90 €
Log in for availability
1 * 5 items
Product Details & Documents
Clips made of POM for conical ground joints.
  • Material does not scratch glass surfaces
  • Resistant to temperatures up to +150 °C
Specifications
  • For NS:
    24/29
  • Colour:
    Green
  • Seq:
    05