Clips for conical ground joints
Blue, 19/26
HECH42386019
Clips made of POM for conical ground joints.
- Material does not scratch glass surfaces
- Resistant to temperatures up to +150 °C
- For NS:19/26
- Colour:Blue
- Seq:04
Clips made of POM for conical ground joints.