Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD UKAS ISO/IEC17025 1 * 500 mL
Catalog # SIALN35-500ML
Supplier:
Merck
Expand image
VISCOSITY STANDARD UKAS ISO/IEC17025 1 * 500 mL
Catalog # SIALN35-500ML
Supplier:
Merck