Optimum Growth Flask
Optimum Growth™ flask, 1,6 L
Supplier: THOMSON INSTRUMENT
Jump to:
Product Details & Documents
Optimum Growth™ flasks (patented) give excellent growth and expression for both mammalian and insect cell lines.
- Baffles designed for high aeration and low shear
- Same footprint as compairable Fernbach flask
- Less foaming than disposable Fernbach flask
- Transfer cap connects directly to Wave Bags™ and bioreactors with quick connect or tube fusing
- 0,2 μm vented cap
- Individually packaged and sterilised
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:1,6 L
- Description:Optimum Growth™ flask
- Seq:4



