Orders LinkContactUsLinkComponent
Optimum Growth Flask
Optimum Growth™ flask, 125 ml
  THMS931110
Optimum Growth Flask
  THMS931110
Optimum Growth Flask
Optimum Growth Flask
Optimum Growth™ flask, 125 ml
566,00 €
1 * 50 items
Optimum Growth™ flasks (patented) give excellent growth and expression for both mammalian and insect cell lines.
  • Baffles designed for high aeration and low shear
  • Same footprint as compairable Fernbach flask
  • Less foaming than disposable Fernbach flask
  • Transfer cap connects directly to Wave Bags™ and bioreactors with quick connect or tube fusing
  • 0,2 μm vented cap
  • Individually packaged and sterilised
  • Capacity:
    125 ml
  • Description:
    Optimum Growth™ flask
  • Seq:
    1