Order Entry
Export
Orders LinkContactUsLinkComponent
Eppendorf® Conical Tubes with SnapTec® Cap

Eppendorf® Conical Tubes with SnapTec® Cap

Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free, Colourless, 150 tubes (6 bags×25 tubes)
Catalog # 525-1605
   Low Manufacturing Impact|Product Transparency and Disclosure
Supplier:   EPPENDORF
Eppendorf® Conical Tubes with SnapTec® Cap
Catalog # 525-1605
Supplier:  EPPENDORF
   Low Manufacturing Impact|Product Transparency and Disclosure
Eppendorf® Conical Tubes with SnapTec® Cap
Eppendorf® Conical Tubes with SnapTec® Cap

Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free, Colourless, 150 tubes (6 bags×25 tubes)

Price
60,50 €
Log in for availability
1 * 150 items
Product Details & Documents

Eppendorf® Conical Tubes with SnapTec® caps offer secure sample containment for diverse applications. The innovative SnapTec® Cap ensures easy, secure sealing, making them ideal for molecular biology, genomics, and more. The conical design aids optimal sample recovery, enhancing efficiency and reliability in the lab.

  • SnapTec® caps allow effortless, secure sealing for sample integrity in molecular biology and genomics
  • Conical design enhances recovery, crucial for applications like PCR, DNA/RNA isolation, and centrifugation
  • Well-suited for various lab tasks, ensuring adaptability across experiments
  • Durable construction crafted for longevity, supporting applications like cell culture, protein purification, and more
  • Streamlines processes, benefitting applications such as qPCR, cloning, and sample storage
  • Available in 25 ml and SnapTec® 50 to 45 ml formats

Eppendorf® Conical Tubes with SnapTec® Caps redefine sample containment, catering to diverse lab applications. The innovative SnapTec® Cap ensures secure sealing for molecular biology and genomics, preserving sample integrity during storage. Versatile across lab tasks, they adapt to cell culture and protein purification. Crafted for durability, these tubes support longevity in various applications. Elevate scientific processes with Eppendorf®, offering trusted quality for versatile lab applications.

Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Material: PP
Maximum RCF: 17000×g

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Sterile Grade can be downloaded from the Eppendorf website.

Specifications
  • Packed:
    150 tubes (6 bags×25 tubes)
  • Colour:
    Colourless
  • Description:
    Sterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free
  • Seq:
    03
  • Cat. No.:
    525-1605
  • Operating temperature range:
    −86...+100 °C
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001, ACT Label
  • Dimensions:
    Ø 30 mm
  • Autoclavable:
    When open, 121 °C, 20 min
  • Environmentally Preferable:
    Low Manufacturing Impact|Product Transparency and Disclosure
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Single tube stand, for 25 and 50 ml tubes
    Single tube stand, for 25 and 50 ml tubes
    EPPE0030119.860
    Log in for availability
    1 * 4 items - 24,50 €

Product Information

  • Catalog #
    Availability
    Price
  • 12 positions, 6 for 5,0 and 15 ml tubes; 6 for 25 and 50 ml tubes
    12 positions, 6 for 5,0 and 15 ml tubes; 6 for 25 and 50 ml tubes
    EPPE0030119.835
    Log in for availability
    1 * 2 items - 52,50 €

Product Information

  • Catalog #
    Availability
    Price
  • Tube holder 25/50 ml, for 4×25/50 ml conical tubes
    Tube holder 25/50 ml, for 4×25/50 ml conical tubes
    EPPE5353040.156
    Log in for availability
    1 * 1 items - 114,00 €

Product Information

  • Catalog #
    Compartments
    For
    Availability
    Price
  • Hole Ø: 30,3 mm
    Hole Ø: 30,3 mm
    EPPE0030140.729
    3×3
    Tubes 25 ml
    Log in for availability
    1 * 2 items - 23,80 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter, for 1 Eppendorf conical tube 25 ml with snap cap 25 ml snap cap, 6 peices.
    Adapter, for 1 Eppendorf conical tube 25 ml with snap cap 25 ml snap cap, 6 peices.
    EPPE5820733.004
    Log in for availability
    1 * 6 items - 34,70 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter 25 ml for SmartBlock 50 ml - set consisting of 4 adapters and removal tool
    Adapter 25 ml for SmartBlock 50 ml - set consisting of 4 adapters and removal tool
    EPPE5365000.109
    Log in for availability
    1 * 1 KIT - 176,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Adapter, for 1 Eppendorf conical tube 25 ml with screw cap 25 ml screw cap, 6 pc
    Adapter, for 1 Eppendorf conical tube 25 ml with screw cap 25 ml screw cap, 6 pc
    EPPE5820734.000
    Log in for availability
    1 * 6 items - 35,30 €