Eppendorf® Conical Tubes with SnapTec® Cap
Product Details & Documents
Eppendorf® Conical Tubes with SnapTec® caps offer secure sample containment for diverse applications. The innovative SnapTec® Cap ensures easy, secure sealing, making them ideal for molecular biology, genomics, and more. The conical design aids optimal sample recovery, enhancing efficiency and reliability in the lab.
- SnapTec® caps allow effortless, secure sealing for sample integrity in molecular biology and genomics
- Conical design enhances recovery, crucial for applications like PCR, DNA/RNA isolation, and centrifugation
- Well-suited for various lab tasks, ensuring adaptability across experiments
- Durable construction crafted for longevity, supporting applications like cell culture, protein purification, and more
- Streamlines processes, benefitting applications such as qPCR, cloning, and sample storage
- Available in 25 ml and SnapTec® 50 to 45 ml formats
Eppendorf® Conical Tubes with SnapTec® Caps redefine sample containment, catering to diverse lab applications. The innovative SnapTec® Cap ensures secure sealing for molecular biology and genomics, preserving sample integrity during storage. Versatile across lab tasks, they adapt to cell culture and protein purification. Crafted for durability, these tubes support longevity in various applications. Elevate scientific processes with Eppendorf®, offering trusted quality for versatile lab applications.
Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.
Material: PP
Maximum RCF: 17000×g
: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Sterile Grade can be downloaded from the Eppendorf website.
Specifications
- Packed:200 tubes (4 bags×50 tubes), 4 single tube stands, 6 adapters for rotors with bore holes for 50 ml conical tubes
- Colour:Colourless
- Description:Starter pack, PCR clean (non sterile)
- Seq:05
- Cat. No.:525-1607
- Operating temperature range:−86...+100 °C
- Environmental certification:Renewable Energy Certificate, ISO 14001, ACT Label
- Dimensions:Ø 30 mm
- Autoclavable:When open, 121 °C, 20 min
- Environmentally Preferable:Low Manufacturing Impact|Product Transparency and Disclosure
Accessories
Product Information
- Catalog #AvailabilityPrice
- Single tube stand, for 25 and 50 ml tubesSingle tube stand, for 25 and 50 ml tubesEPPE0030119.860Log in for availability1 * 4 items - 24,50 €
Product Information
- Catalog #AvailabilityPrice
- 12 positions, 6 for 5,0 and 15 ml tubes; 6 for 25 and 50 ml tubes12 positions, 6 for 5,0 and 15 ml tubes; 6 for 25 and 50 ml tubesEPPE0030119.835Log in for availability1 * 2 items - 52,50 €
Product Information
- Catalog #AvailabilityPrice
- Tube holder 25/50 ml, for 4×25/50 ml conical tubesTube holder 25/50 ml, for 4×25/50 ml conical tubesEPPE5353040.156Log in for availability1 * 1 items - 114,00 €
Product Information
- Catalog #CompartmentsForAvailabilityPrice
- Hole Ø: 30,3 mm
Product Information
- Catalog #AvailabilityPrice
- Adapter, for 1 Eppendorf conical tube 25 ml with snap cap 25 ml snap cap, 6 peices.Adapter, for 1 Eppendorf conical tube 25 ml with snap cap 25 ml snap cap, 6 peices.EPPE5820733.004Log in for availability1 * 6 items - 34,70 €
Product Information
- Catalog #AvailabilityPrice
- Adapter 25 ml for SmartBlock 50 ml - set consisting of 4 adapters and removal toolAdapter 25 ml for SmartBlock 50 ml - set consisting of 4 adapters and removal toolEPPE5365000.109Log in for availability1 * 1 KIT - 176,00 €
Product Information
- Catalog #AvailabilityPrice
- Adapter, for 1 Eppendorf conical tube 25 ml with screw cap 25 ml screw cap, 6 pcAdapter, for 1 Eppendorf conical tube 25 ml with screw cap 25 ml screw cap, 6 pcEPPE5820734.000Log in for availability1 * 6 items - 35,30 €



