Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Grace Vision HT, C18-HL, 1.5 um, 2.0x100 mm column 1 * 1 items
Catalog # MAIS5142547
Supplier:
DR MAISCH
Expand image
Grace Vision HT, C18-HL, 1.5 um, 2.0x100 mm column 1 * 1 items
Catalog # MAIS5142547
Supplier:
DR MAISCH