Orders LinkContactUsLinkComponent
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
P8x10, 1 - 10 µl, ±0,08 - ±0,20 µl, ≤ 0,05 - ≤ 0,10 µl, Red, D10 / DL10 / DF10 / DFL10
  613-7152
 :   GILSON
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
  613-7152
 :  GILSON
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
Multi-channel pipettes, mechanical, variable volume, PIPETMAN®
P8x10, 1 - 10 µl, ±0,08 - ±0,20 µl, ≤ 0,05 - ≤ 0,10 µl, Red, D10 / DL10 / DF10 / DFL10
801,00 €
1 * 1 items
PIPETMAN®, recognized as the pipetting standard for its reliability and proven durability, is a fully adjustable, air-displacement pipette that uses disposable tips. PIPETMAN combines legendary accuracy, precision, and durability with easy pipetting and handling. The pipettes come in a large range of volumes with reduced pipetting forces for hours of comfortable pipetting.
  • Legendary performance
  • Easy pipetting
  • Built-to-last and convenient
  • Simple to maintain
Accurate and precise, PIPETMAN air-displacement pipettes are recognized as one of the world standard. They come in a large range of volumes, including single and multichannel models, allowing you to handle various sample types with confidence. PIPETMAN feels like PIPETMAN® Classic but with lower forces for enhanced pipetting habits with a standard pipette. Made of PVDF and stainless steel, PIPETMAN offers premium durability. Built-to-last, it is the economical solution for years of pipetting. Routine cleaning and maintenance are all that is required to keep PIPETMAN in top condition for years. The PIPETMAN design allows for easy access to user-serviceable parts, along with a fully autoclavable tip holder and ejector.
 : Models equipped with a tip ejector are only sold with a stainless steel tip ejector. Plastic tip ejectors are available in the accessories.
  • Colour code:
    Red
  • Volume:
    1 - 10 µl
  • Description:
    P8x10
  • Seq:
    01
  • Accuracy:
    ±0,08 - ±0,20 µl
  • Imprecision:
    ≤ 0,05 - ≤ 0,10 µl
  • Recommended tips:
    D10 / DL10 / DF10 / DFL10
Accessories
  •  
    Description
    Colour
  • 612-1816
    Rotating pipette stand, CARROUSEL™, for 7 Gilson pipettes (except Pipetman Concept®)
    White
    1 * 1 items - 126,00 €
  •  
    Description
    Colour
  • GILSF161405
    Angled pipette stand, TRIO™, for 3 Gilson pipettes (except Pipetman Concept®)
    Translucent
    1 * 1 items - 111,00 €
  •  
    Description
    Colour
  • GILSF161406
    Pipette holder, SINGLE™, for one Gilson pipette, can be stucked to any smooth, dry surface or fastened with screws
    White
    1 * 1 items - 19,00 €
  •  
    Description
  • GILSF107028
    Plastic ejector for P20
    1 * 1 items - 11,60 €
  •  
    Description
  • GILSF107027
    Plastic ejector for P2/P10 with adapter
    1 * 1 items - 15,80 €
  •  
    Description
  • GILSF107031
    Plastic ejector for P1000
    1 * 1 items - 11,60 €
  •  
    Description
  • GILSF107030
    Plastic ejector for P200
    1 * 1 items - 11,60 €
  •  
    Description
  • GILSF107029
    Plastic ejector for P100
    1 * 1 items - 11,60 €