High-Performance Chromatography Refrigerators, TSX
Product Details & Documents
The TSX high-performance chromatography refrigerators are designed for a variety of applications requiring close temperature control, full access to chromatography instrumentation, and easy set-up of instrumentation and apparatus within the chamber. The TSX chromatography series features industry-leading safety features to ensure maximum protection.
- Forced-air circulation for temperature uniformity at all shelf levels and Heat-free defrost for maximum temperature uniformity
- Quiet operation at just 52 dBA
- Two 50 mm side wall access ports with insulated hinged covers
- Four 50,8 mm casters for easy mobility; the front two are lockable
- Bright, LED interior lighting
- Keyed on/off switch with easy to use, set-point security
- Triple pane glass doors and self-closing door with 90° stop to assist with inventory loads
- Safety switch protected internal electrical outlet on all models (Duplex outleton A and D models, single outlet on V models)
- Energy Star certified and SNAP compliant
- DeviceLink compatible
TSX Series high-performance chromatography refrigerators-lab and clinical-grade storage that adapts to you and your environment. Temperature variation can have an impact on the viability and efficacy of vaccines, medication, reagents and other temperature-sensitive materials. These variations can shorten product shelf life, degrade enzymes and reduce vaccine effectiveness. Therefore, it is important to choose a high-performance refrigerator or freezer that has been designed for the demanding standards of the laboratory and clinical environment.
The sustainable, SNAP compliant choice - Thermo Scientific™ TSX Series high-performance refrigerators and freezers are designed with features that support sample protection, sustainability objectives and the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP). Our V-drive technology is designed to provide temperature uniformity that continually adapts to the lab or clinical environment, offering significant energy savings without compromising protection.
Whisper quiet operation - Compared to conventional refrigerant, high-performance models†, the TSX Series offers a whisper quiet operation, so you can bring the refrigerator or freezer out of the hallway and back into your lab without disrupting your audio environment.
Significant energy savings - While conventional refrigerant, high-performance refrigerators and freezers can run at very high kWh/day in energy usage, the TSX Series by comparison has been designed to reduce energy usage without compromising performance.
Note: Dimensions shown below are W×D×H.
Certifications: Meets cULus, CE, UPS 800, ISO Class 6, EPA Energy Star, EPA SNAP.
Specifications
- W×D×H ext.:864×947×1994 mm
- Capacity:827 L
- Description:TSX high performance chromatography refrigerator CEE 7/7 plug
- Seq:02
- W×D×H int.:762×723×1473 mm
- Temperature:2...8 °C
- Cat. No.:HERATSX3005CV
- Environmental certification:Energy Star; EPA SNAP Compliant
- Defrost:Automatic
- Plug:CEE 7/7
- Door type:1 glass swing door
- Electrical:208 - 230 V, 50/60 Hz



