Order Entry
Export
Orders LinkContactUsLinkComponent
Polysorbate 80 FCC

Polysorbate 80 FCC

Glass, amber, 4 L
Catalog # SPCMP1179-4LTGL
Supplier:   Spectrum Chemical
Polysorbate 80 FCC
Catalog # SPCMP1179-4LTGL
Supplier:  Spectrum Chemical
CAS Number:  9005-65-6
Polysorbate 80 FCC

Glass, amber, 4 L

CAS Number:  9005-65-6
Price
449,00 €
Log in for availability
1 * 4 L
Product Details & Documents

Polysorbate 80, FCC is used as a wetting agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Kosher certified

Storage temperature - Keep tightly closed, protect from light.

Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  • Melting Pt:100 °C
  • Flash Pt:113 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:500-019-9
Specifications

Specification Test Results

  • Assay:
    65% Min.

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    4 L

Specifications