Benzalkonium chloride solution 50% NF
500 mL
Supplier: Spectrum Chemical
Benzalkonium chloride solution 50% NF
Catalog # SPCMB1068-500MLGL
Supplier: Spectrum Chemical
CAS Number: 63449-41-2
Jump to:
Product Details & Documents
Benzalkonium Chloride Solution, 50 Percent, NF is used in the pharmaceutical industry as an antimicrobial preservative, antiseptic, disinfectant, solubilising agent, and wetting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
- CAS Number:63449-41-2
- Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
- MDL Number:MFCD00145757
- Melting Pt:29…34 °C
- Density:0,98 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:264-151-6
- UN:2920
- ADR:8.3,II
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Assay (%):47.5
Specifications
- Pk:500 mL



