Order Entry
Export
Orders LinkContactUsLinkComponent
Benzalkonium chloride solution 50% NF

Benzalkonium chloride solution 50% NF

Glass, amber, 4 L
Catalog # SPCMB1068-4LTGL
Corrosive
Irritant
Environment
Supplier:   Spectrum Chemical
Benzalkonium chloride solution 50% NF
Catalog # SPCMB1068-4LTGL
Supplier:  Spectrum Chemical
CAS Number:  63449-41-2
Benzalkonium chloride solution 50% NF

Glass, amber, 4 L

CAS Number:  63449-41-2
Price
797,00 €
Log in for availability
1 * 4 L
Product Details & Documents

Benzalkonium Chloride Solution, 50 Percent, NF is used in the pharmaceutical industry as an antimicrobial preservative, antiseptic, disinfectant, solubilising agent, and wetting agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride
  • CAS Number:63449-41-2
  • Formula:C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
  • MDL Number:MFCD00145757
  • Melting Pt:29…34 °C
  • Density:0,98 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:264-151-6
  • UN:2920
  • ADR:8.3,II

Documents

Specifications

Specification Test Results

  • Assay (%):
    47.5

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    4 L

Specifications