0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Colour Standard Gardner 4 1 * 100 mL
REGEGARD041
:
REAGECON
Colour Standard Gardner 4 1 * 100 mL
REGEGARD041
:
REAGECON