Spare keys for stopcocks, Pyrex®
Replacement keys, for 18,8 mm Ø, 6,0 mm, SSD series stopcocks
Catalog # BIBBSSD106K
Supplier: SciLabware
Jump to:
Product Details & Documents
Spare keys for stopckocks, made of borosilicate glass.
- Straight bore
- Interchangeable
: Supplied complete with key retaining device.
Specifications
- For:SSD series stopcocks
- Hole Ø:6,0 mm
- Description:Replacement keys, for 18,8 mm Ø
- Seq:04



