Orders LinkContactUsLinkComponent
Accessories for PLATEMASTER®
Accessories for PLATEMASTER®
384-well plate adapter, positioning by wheel
Catalog # 613-5910
Supplier:   GILSON
Accessories for PLATEMASTER®
Catalog # 613-5910
Supplier:  GILSON
Accessories for PLATEMASTER®
Accessories for PLATEMASTER®
384-well plate adapter, positioning by wheel
Price
1 664,00 €
Log in for availability
1 * 1 items
Product Details & Documents
The Pipetting Head Height Adjuster is an accessory for the PLATEMASTER® that allows for easy altering of pipetting height, ensuring appropriate tip depth within shallow- or deep-well microplates.
The Lubrication Box of O-rings is an accessory for the PLATEMASTER®. We recommend lightly lubricating the external O-ring between regular maintenance intervals in situations of intensive use and to avoid ill-fitting tips.
Specifications
  • Description:
    384-well plate adapter, positioning by wheel