ContactUsLinkComponent
Stericup Quick Release-GV Sterile Vacuum Filtration System 1000 mL process volume, 0.22 µm pore size, PVDF membrane, pkg of 12, radio-sterilized 1 * 12 items
Catalog # MILFS2GVU10R
Supplier:  Merck
Stericup Quick Release-GV Sterile Vacuum Filtration System 1000 mL process volume, 0.22 µm pore size, PVDF membrane, pkg of 12, radio-sterilized 1 * 12 items
Catalog # MILFS2GVU10R
Supplier:  Merck