Order Entry
Export
Orders LinkContactUsLinkComponent
Mini coolers, True North®

Mini coolers, True North®

True North® mini-cooler, –20 °C, clear lid, blue, –15 °C for 3,5 h temperature retention, 32×0,5 - 2 ml, 4×8 array, 245×105×100 mm
Catalog # HEATHS120074
Supplier:   HEATHROW
Mini coolers, True North®
Catalog # HEATHS120074
Supplier:  HEATHROW
Mini coolers, True North®
Mini coolers, True North®

True North® mini-cooler, –20 °C, clear lid, blue, –15 °C for 3,5 h temperature retention, 32×0,5 - 2 ml, 4×8 array, 245×105×100 mm

Price
502,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Protect temperature-sensitive samples at the bench or in the freezer.

  • Polycarbonate construction with rubber non-slip feet
  • Gridded and keyed one-position lids for easy sample identification

Ideal for keeping PCR® reagents, enzymes, and other biochemical reagents cool while working at the bench.
Filled with insulating gel, the mini coolers can also be used for controlled sample freezing and for longer temperature retention samples.

Coolers HEATHS120072 and HEATHS120075 are filled with a non-toxic self-insulating gel.

Ordering information: Versions available for 0,5 and 1,5/2,0 ml microcentrifuge tubes and cryogenic vials and a 96-place version for 0,2 ml tubes and assay plates.

Delivery information: Supplied with 0,5 ml tube adapters as standard and temperature-retention graph.

Specifications
  • Capacity:
    32×0,5 - 2 ml, 4×8 array
  • Description:
    True North® mini-cooler, –20 °C, clear lid, blue, –15 °C for 3,5 h temperature retention
  • Dimensions:
    245×105×100 mm
  • Seq:
    05