Mini coolers, True North®
True North® mini-cooler, –20 °C, clear lid, blue, –15 °C for 2,5 h temperature retention, 12×0,5 - 2 ml, 3×4 array, 145×105×100 mm
Catalog # HEATHS120073
Supplier: HEATHROW
Jump to:
Product Details & Documents
Protect temperature-sensitive samples at the bench or in the freezer.
- Polycarbonate construction with rubber non-slip feet
- Gridded and keyed one-position lids for easy sample identification
Ideal for keeping PCR® reagents, enzymes, and other biochemical reagents cool while working at the bench.
Filled with insulating gel, the mini coolers can also be used for controlled sample freezing and for longer temperature retention samples.
Coolers HEATHS120072 and HEATHS120075 are filled with a non-toxic self-insulating gel.
: Versions available for 0,5 and 1,5/2,0 ml microcentrifuge tubes and cryogenic vials and a 96-place version for 0,2 ml tubes and assay plates.
: Supplied with 0,5 ml tube adapters as standard and temperature-retention graph.
Specifications
- Capacity:12×0,5 - 2 ml, 3×4 array
- Description:True North® mini-cooler, –20 °C, clear lid, blue, –15 °C for 2,5 h temperature retention
- Dimensions:145×105×100 mm
- Seq:02



