Order Entry
Export
Orders LinkContactUsLinkComponent
D-(-)-Mannitol ≥96%, powder USP

D-(-)-Mannitol ≥96%, powder USP

Poly bottle, 500 g
Catalog # SPCMMA165-500GM
Supplier:   Spectrum Chemical
D-(-)-Mannitol ≥96%, powder USP
Catalog # SPCMMA165-500GM
Supplier:  Spectrum Chemical
CAS Number:  69-65-8
D-(-)-Mannitol ≥96%, powder USP
D-(-)-Mannitol ≥96%, powder USP

Poly bottle, 500 g

CAS Number:  69-65-8
Price
417,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Mannitol, Powder, USP.

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1,49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8

Documents

Specifications

Specification Test Results

  • Assay:
    96% Min.

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g

Specifications