Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Peracetic Acid Chemkeys, range 0.04 - 50.0 mg/l PAA., (Box of 25) 1 * 25 items
HACH8635200
:
Hach
Peracetic Acid Chemkeys, range 0.04 - 50.0 mg/l PAA., (Box of 25) 1 * 25 items
HACH8635200
:
Hach