ContactUsLinkComponent
ChromegaChiral(TM) Chiral CCO F4, 10 Micron HPLC Column, 10 mum particle size, L x I.D. 25 cm x 20 mm 1 * 1 items
Catalog # SIALES5604
Supplier:  Merck
ChromegaChiral(TM) Chiral CCO F4, 10 Micron HPLC Column, 10 mum particle size, L x I.D. 25 cm x 20 mm 1 * 1 items
Catalog # SIALES5604
Supplier:  Merck