Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
ChromegaChiral(TM) Chiral CC4, 10 Micron HPLC Column, 10 mum particle size, L x I.D. 10 cm x 3.0 mm 1 * 1 items
SIALES5727
:
Merck
ChromegaChiral(TM) Chiral CC4, 10 Micron HPLC Column, 10 mum particle size, L x I.D. 10 cm x 3.0 mm 1 * 1 items
SIALES5727
:
Merck