ContactUsLinkComponent
ChromegaChiral(TM) Chiral CCO F4, 3 Micron HPLC Column, 3 mum particle size, L x I.D. 10 cm x 4.6 mm 1 * 1 items
Catalog # SIALES5578
Supplier:  Merck
ChromegaChiral(TM) Chiral CCO F4, 3 Micron HPLC Column, 3 mum particle size, L x I.D. 10 cm x 4.6 mm 1 * 1 items
Catalog # SIALES5578
Supplier:  Merck