ContactUsLinkComponent
ChromegaChiral(TM) Chiral CCO F4, 3 Micron HPLC Column, 3 mum particle size, L x I.D. 5 cm x 2.1 mm 1 * 1 items
Catalog # SIALES5573
Supplier:  Merck
ChromegaChiral(TM) Chiral CCO F4, 3 Micron HPLC Column, 3 mum particle size, L x I.D. 5 cm x 2.1 mm 1 * 1 items
Catalog # SIALES5573
Supplier:  Merck