ContactUsLinkComponent
ChromegaChiral(TM) Chiral CCA, 5 Micron HPLC Column, 5 mum particle size, L x I.D. 25 cm x 3.0 mm 1 * 1 items
Catalog # SIALES5436
Supplier:  Merck
ChromegaChiral(TM) Chiral CCA, 5 Micron HPLC Column, 5 mum particle size, L x I.D. 25 cm x 3.0 mm 1 * 1 items
Catalog # SIALES5436
Supplier:  Merck