Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Rabbit monoclonal antibody raised against synthetic peptide of human VSV-G-Tag. 1 * 100 µG
Catalog # ABNOMAB20851
Supplier:
Abnova
Expand image
Rabbit monoclonal antibody raised against synthetic peptide of human VSV-G-Tag. 1 * 100 µG
Catalog # ABNOMAB20851
Supplier:
Abnova