Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
TBUXPHOS-PD-G
TBUXPHOS-PD-G
Print
TBUXPHOS-PD-G
Catalog # COBBQM-4697-5G
Supplier:
COMBI-BLOCKS
TBUXPHOS-PD-G
Catalog # COBBQM-4697-5G
Supplier:
COMBI-BLOCKS
CAS Number:
Expand image
Return to Top
TBUXPHOS-PD-G
Add to cart
CAS Number:
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 5 g
-
+
Inquire For Price
Return Policy
Product Details & Documents