Orders LinkContactUsLinkComponent
Bumper Guards
Bumper Guards
Bumper large rectangle surface, 75×20×1200 mm
Catalog # ERGOLRSB120-BK
Supplier:   ERGOMAT
Bumper Guards
Catalog # ERGOLRSB120-BK
Supplier:  ERGOMAT
Bumper Guards
Bumper Guards
Bumper large rectangle surface, 75×20×1200 mm
Price
33,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These Ergomat bumper guards, cart stops and edge protectors offer a variety of devices to prevent personal injury or product damage in a dynamic work environment.
  • Resistant polyurethane foam with self-adhesive attachment to rack beams, posts and uprights
  • Use anywhere workers might run into objects - supports, stands, low hanging objects
  • Round, flat, and edge surface protection
  • Yellow and black hazard striped
These guards prevent cuts and bruises from sharp edges with edge protecting bumper guards that slide over the sharp edge and hold themselves in place. A cushion of foam rubber helps soften the blow and takes the bite out of sharp edges. They also protect workers as they move in and out of rack space with bumper guards on the bottom of rack beams and sharp edges.
They help to prevent bumps, bruises and cuts by providing a soft cushion where workers might bump their heads and shoulders. They also help to reduce damage from light impact to walls and equipment with flat foam rubber bumper guards placed where impact is likely. For heavier impact, select a steel supported bumper guard where car doors might hit a wall or other similar impacts.
Certifications: DIN 53505, EN ISO 14664-1, meets Cleanroom ISO Class 5 (Class 100) compliance.
Specifications
  • Description:
    Bumper large rectangle surface
  • Dimensions:
    75×20×1200 mm