Isofreeze® racks with temperature indicator
Isofreeze MCT rack, for 1,5 and 2,0 ml micro tubes and screw cap micro tubes, 24 (4×6), 305×305×298 mm
Catalog # SARS95.983
Supplier: SARSTEDT
Jump to:
Product Details & Documents
Isofreeze® racks are for use as a pipetting and storage station for temperature sensitive applications like enzyme analyses, PCR or cell based assays. They prevent sample damage caused by premature warming.
- Transparent cap with white frame
- Cooling block undergoes a clearly visible colour change from purple to pink when product leaves optimal temperature range (at 7 °C)
- With the lid closed, the sample temperature can be maintained at ~4 °C for three hours
Specifications
- No. of Holes:24 (4×6)
- Description:Isofreeze MCT rack, for 1,5 and 2,0 ml micro tubes and screw cap micro tubes
- Seq:1
- W×D×H:305×305×298 mm



