Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REPROSIL FLUOSIL 100-PFP. 3µM, 150X4,6MM 1 * 1 items
Catalog # MAISFL15.PFP.S1546
Supplier:
DR MAISCH
Expand image
REPROSIL FLUOSIL 100-PFP. 3µM, 150X4,6MM 1 * 1 items
Catalog # MAISFL15.PFP.S1546
Supplier:
DR MAISCH