Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PROSPHERE 300 C4 5 µM 150 X 4.6 MM 1 * 1 items
Catalog # MAIS33989
Supplier:
DR MAISCH
Expand image
PROSPHERE 300 C4 5 µM 150 X 4.6 MM 1 * 1 items
Catalog # MAIS33989
Supplier:
DR MAISCH