Order Entry
Export
Orders LinkContactUsLinkComponent
 

HEATING CIRCULATOR TERMOTRONIC 100 1 * 1 items

  SELE3000540
 :   SELECTA
HEATING CIRCULATOR TERMOTRONIC 100 1 * 1 items
  SELE3000540
 :  SELECTA
HEATING CIRCULATOR TERMOTRONIC 100 1 * 1 items
 
719,75 €
   
1 * 1 items
 
 
  • Storage Conditions:
    2…8 °C
  • Cat. No.:
    78458-583
  • M-Clarity quality level:
    MQ100
  • Type:
    Certified reference material
  • Flash Point:
    17.0 °C - closed cup
  • Hazard Class Description:
    H225 - H302 + H312 + H332 - H315 - H319
  • Shipping Conditions:
    WICE
  • CAS No.:
    111-03-5