Order Entry
Export
Orders LinkContactUsLinkComponent
POTASSIUM HYDROXIDE STD SOLUTION 12N 1 * 500 mL

POTASSIUM HYDROXIDE STD SOLUTION 12N 1 * 500 mL

Catalog # HACH23049H
Supplier:   Hach
POTASSIUM HYDROXIDE STD SOLUTION 12N 1 * 500 mL
Catalog # HACH23049H
Supplier:  Hach
CAS Number:  
POTASSIUM HYDROXIDE STD SOLUTION 12N 1 * 500 mL
CAS Number:  
Price
60,60 €
Log in for availability
1 * 500 mL
Product Details & Documents