Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DEVELOSIL TMS-UG 5µ, 50 X 4.6MM 1 * 1 items
Catalog # HISMUG1546050W
Supplier:
HICHROM
Expand image
DEVELOSIL TMS-UG 5µ, 50 X 4.6MM 1 * 1 items
Catalog # HISMUG1546050W
Supplier:
HICHROM