Orders LinkContactUsLinkComponent
Ready-to-use tyramide amplification buffer, 1X
Ready-to-use tyramide amplification buffer, 1X
Ready-to-use tyramide amplification buffer, 1X, 100 mL
Catalog # 22027-100ML
Supplier:   Biotium
Ready-to-use tyramide amplification buffer, 1X
Catalog # 22027-100ML
Supplier:  Biotium
CAS Number:  
Ready-to-use tyramide amplification buffer, 1X
Ready-to-use tyramide amplification buffer, 1X
Ready-to-use tyramide amplification buffer, 1X, 100 mL
CAS Number:  
Price
698,00 €
Log in for availability
1 * 100 mL
Product Details & Documents
For simple, optimised tyramide amplification of immunostaining in cells or tissue sections. No need to add hydrogen peroxide before use.
  • Ready-to-use
  • Simply dilute tyramide probe in buffer and proceed to signal development
  • Compatible with tyramides from Biotium or other suppliers
The Tyramide Amplification Buffer Plus (BTIU22029), a improved buffer for tyramide signal amplification provides significantly improved brightness, specificity, and sensitivity over this original buffer.
Simply mix the tyramide compound with the buffer to make a working solution for signal amplification. Although specifically optimised for our CF® Dye-, Biotin XX-, and DNP-tyramide products, Ready-to-Use Tyramide Amplification Buffer is also compatible with tyramide probes from other manufacturers.
Documents
Specifications
Specifications
  • Pk:
    100 mL
  • Description:
    Ready-to-use tyramide amplification buffer, 1X
Specifications