Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ECONO-CAP EC-WAX+, 30 M X 0.32 MM X 0.25 1 * 1 items
Catalog # HISM8635053
Supplier:
HICHROM
Expand image
ECONO-CAP EC-WAX+, 30 M X 0.32 MM X 0.25 1 * 1 items
Catalog # HISM8635053
Supplier:
HICHROM