Orders LinkContactUsLinkComponent
Accessories for Eppendorf microcentrifuge
Accessories for Eppendorf microcentrifuge
Adapters for rotors, for 3 oak ridge tubes 30 ml, for rotor FA-6x250, 2 pieces
Catalog # 521-2697
Supplier:   EPPENDORF
Accessories for Eppendorf microcentrifuge
Catalog # 521-2697
Supplier:  EPPENDORF
Accessories for Eppendorf microcentrifuge
Accessories for Eppendorf microcentrifuge
Adapters for rotors, for 3 oak ridge tubes 30 ml, for rotor FA-6x250, 2 pieces
Price
335,00 €
Log in for availability
1 * 2 items
Product Details & Documents
Specifications
  • Description:
    Adapters for rotors, for 3 oak ridge tubes 30 ml, for rotor FA-6x250, 2 pieces
  • Seq:
    22