Orders LinkContactUsLinkComponent
Accessories for Eppendorf microcentrifuge
Accessories for Eppendorf microcentrifuge
Adapters for rotors, for 50 ml conical skirted tubes 50 ml, 2 pieces
Catalog # 521-2696
Supplier:   EPPENDORF
Accessories for Eppendorf microcentrifuge
Catalog # 521-2696
Supplier:  EPPENDORF
Accessories for Eppendorf microcentrifuge
Accessories for Eppendorf microcentrifuge
Adapters for rotors, for 50 ml conical skirted tubes 50 ml, 2 pieces
Price
319,00 €
Log in for availability
1 * 2 items
Product Details & Documents
Specifications
  • Description:
    Adapters for rotors, for 50 ml conical skirted tubes 50 ml, 2 pieces