Caps for sample tubes, tamper evident
Screw cap, for sample tubes, 30 ml, White
Catalog # SIMPT552-30W
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Caps made of PE feature a long skirt and a super fast thread design allowing them to be removed or sealed with a single turn. Caps are designed with flexible sealing lip which provides leakproof seal.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- For tubes:30 ml
- Colour:White
- Description:Screw cap, for sample tubes
- Seq:07