ContactUsLinkComponent
Thermometer, solid stem, similar to ASTM 9 C,white backed, -5+110:0,5C,red special liquid, 57mm immersion, c.290x6-7mm long, durabl e pigment 1 * 1 items
Catalog # AMARA300150FLDKD4
Supplier:  Amarell
Thermometer, solid stem, similar to ASTM 9 C,white backed, -5+110:0,5C,red special liquid, 57mm immersion, c.290x6-7mm long, durabl e pigment 1 * 1 items
Catalog # AMARA300150FLDKD4
Supplier:  Amarell