Order Entry
Export
Orders LinkContactUsLinkComponent
 

Graphic mats

LogoMat, smooth, 900×600 mm, 11 mm
  ERGOLS6090
 :   ERGOMAT
Graphic mats
  ERGOLS6090
 :  ERGOMAT
Graphic mats
Graphic mats

LogoMat, smooth, 900×600 mm, 11 mm

 
127,00 €
   
1 * 1 items
 

These graphic logo mats are extremely strong and durable while being easy to clean and broadly resistant to incidental liquid spills. These mats are promoting a brand and decorate a space with a functional, practical health and safety product that delivers years of relief and aesthetic enjoyment.

  • Free of silicone and latex
  • Moulded in durable, lightweight polyurethane
  • Can be customised with logos or other desired graphics

Available with ergonomic bubble profile or with moderately textured profile and smooth logo: Infinity bubble logo mat is designed to allow workers an anti-fatigue, ergonomic work station. Smooth logo mat offers a flat surface to accommodate pivoting workers and allow trolley accessibility.

 : Rated to ISO Cleanroom Class 5 (Class 100).

 
  • Description:
    LogoMat, smooth
  • L×W:
    900×600 mm
  • Seq:
    02
  • Thickness:
    11 mm