Cell culture inserts, 2 in 1, cellGrade™ plus
6-well insert 2 in 1, PC, 48×1, 0,4 μm
Catalog # 734-3283
Supplier: Brand
Jump to:
Product Details & Documents
Cell culture inserts with cellGrade™ plus treated membranes for improved cell attachment and growth.
- Can be used as a hanging insert (fits nearly all multiwell plates) or, for bigger wells and cuture dishes, as a standing insert
- cellGrade™ plus surface for excellent cell attachment and growth
- Highly transparent membranes give good visibility of cell cultures
- High pore density contributes to good permeability
- Innovative packaging provides for efficient unpacking and reduces waste
Ordering information: Inserts are available for 6-, 12- and 24-well plate sizes and in two pore sizes (0,4 μm and 8,0 μm) with PC or PET membrane.
Packaging: Individually packed in single blisters or in a plate-style blister.
Specifications
- Membrane:PC
- Packed:48×1
- Pore size:0,4 μm
- Description:6-well insert 2 in 1
- Seq:17



