Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Pellicon XL Ultrafiltration Module Biomax 50 kDa 0.005 m2 1 * 1 ST
Catalog # MILXPXB050A50
Supplier:
Merck
Expand image
Pellicon XL Ultrafiltration Module Biomax 50 kDa 0.005 m2 1 * 1 ST
Catalog # MILXPXB050A50
Supplier:
Merck