Calcium acetate USP
100 g
Catalog # SPCMC1067-100GM
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Synonyms: Calcium acetate, United States Pharmacopeia (USP) Reference Standard, Calcium di(acetate), Acetic acid calcium salt
- CAS Number:62-54-4
- Formula:(H₃CCOO)₂Ca
- MDL Number:MFCD00150019
- Molecular Weight:158.17 g/mol
- Merck Index:12,01683
- Density:1.509 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-540-9
Specifications
Specification Test Results
- Assay:99% Min.
Specifications
- Pk:100 g



