Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
IGMF 30934 Induktiver Sensor M18x1x80, 1.4571/PTFE, Sn 5b, 24VDC 2-Leiter Schließer, -25°C…+120°C, 3 m FEP - Kabel 1 * 1 ST
Catalog # EGEES30934/3M
Supplier:
EGE ELKTRONIK SPEZIAL SENSOREN
Expand image
IGMF 30934 Induktiver Sensor M18x1x80, 1.4571/PTFE, Sn 5b, 24VDC 2-Leiter Schließer, -25°C…+120°C, 3 m FEP - Kabel 1 * 1 ST
Catalog # EGEES30934/3M
Supplier:
EGE ELKTRONIK SPEZIAL SENSOREN