Order Entry
Export
Orders LinkContactUsLinkComponent
Peltor™ Rechargeable Batteries

Peltor™ Rechargeable Batteries

Rechargeable NimH battery pack, 2,4 V, 2500 mAh
Catalog # MMMAACK053
Supplier:   3M
Peltor™ Rechargeable Batteries
Catalog # MMMAACK053
Supplier:  3M
Peltor™ Rechargeable Batteries
Peltor™ Rechargeable Batteries

Rechargeable NimH battery pack, 2,4 V, 2500 mAh

Price
86,60 €
Log in for availability
1 * 1 items
Product Details & Documents

Rechargeable battery pack for use with specific 3M™ Peltor™ headsets.

  • Rechargeable battery pack
  • NiMH, AA, and Lithium-ion packs available
Specifications
  • Description:
    Rechargeable NimH battery pack, 2,4 V, 2500 mAh
  • Seq:
    02