Order Entry
Export
Orders LinkContactUsLinkComponent
Assistent® Brushes with Cotton Tip Bristles

Assistent® Brushes with Cotton Tip Bristles

Cotton, 15 mm, Wire, 250 mm
  HECH91120000002
Assistent® Brushes with Cotton Tip Bristles
  HECH91120000002
Assistent® Brushes with Cotton Tip Bristles
Assistent® Brushes with Cotton Tip Bristles

Cotton, 15 mm, Wire, 250 mm

 
130,00 €
   
1 * 50 items
 

Brushes, light bristles, with cotton tip.

 
  • Bristle material:
    Cotton
  • Brush head Ø:
    15 mm
  • Handle material:
    Wire
  • Seq:
    07
  • Total length:
    250 mm