Labels for BradyJet J2000 Lab printer
Ink pads, J20-PAD, Inkjet, White, J2000, 127×3 mm
Catalog # BRDY150163
Supplier: Brady
Jump to:
Product Details & Documents
These PP BradyJet J2000 Lab printer labels come in matching sizes and shapes for most sample containers including tubes, vials, cryo containers, dishes, plates and general glassware.
- Designed to stay attached and remain legible
- Resist liquid nitrogen, freezers, hot water baths and autoclaves
They can also resist common laboratory chemicals like acetone, ethanol, toluene, xylene and IPA.
Documents
Specifications
- Printer model:J2000
- Printing method:Inkjet
- Colour:White
- Description:Ink pads
- Dimensions:127×3 mm
- Seq:22
- Model:J20-PAD